5-Chlor-2-methyl-4-thiocyanatoanilin 97%

Lieferant: Apollo Scientific

OR909171-1G OR909171-25G OR909171-5G
APOSOR909171-1GEA 109 EUR
APOSOR909171-1G APOSOR909171-25G APOSOR909171-5G
5-Chlor-2-methyl-4-thiocyanatoanilin 97%
5-Chlor-2-methyl-4-thiocyanatoanilin
Formel: C₈H₇ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD12026046
CAS-Nummer: 23530-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR