5-Isopropyl-1H-pyrazole 98%
Lieferant: Apollo Scientific
OR909285-5G
APOSOR909285-5GEA
102
EUR
APOSOR909285-5G
5-Isopropyl-1H-pyrazole 98%
5-Isopropyl-1H-pyrazole
Formel:
C₆H₁₀N₂ Molecular Weight: 110,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16556133 CAS-Nummer: 49633-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...