2-(4-Nitrophenyl)oxazol 98%

Lieferant: Apollo Scientific

OR909356-1G OR909356-5G
APOSOR909356-1GEA 203 EUR
APOSOR909356-1G APOSOR909356-5G
2-(4-Nitrophenyl)oxazol 98%
2-(4-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08276758
CAS-Nummer: 62882-08-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR