4-Bromo-2-formylbenzonitrile 97%

Lieferant: Apollo Scientific
Achtung

OR909386-1G OR909386-250MG
APOSOR909386-1GEA 56.7 EUR
APOSOR909386-1G APOSOR909386-250MG
4-Bromo-2-formylbenzonitrile 97%
4-Bromo-2-formylbenzonitrile
Formel: C₈H₄BrNO
Molecular Weight: 210,03 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15144645
CAS-Nummer: 713141-12-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR