3-Brom-4-methylbenzaldehyd 98%
Lieferant: Apollo Scientific
OR909388-100G
OR909388-10G
OR909388-25G
APOSOR909388-100GEA
449
EUR
APOSOR909388-100G
APOSOR909388-10G
APOSOR909388-25G
3-Brom-4-methylbenzaldehyd 98%
3-Bromo-4-methylbenzaldehyde
Formel:
C₈H₇BrO Molecular Weight: 199.0500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971197 CAS-Nummer: 36276-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...