3-Ethoxypiperidine 97%

Lieferant: Apollo Scientific

OR909417-1G OR909417-5G
APOSOR909417-1GEA 85.7 EUR
APOSOR909417-1G APOSOR909417-5G
3-Ethoxypiperidine 97%
3-Ethoxypiperidine
Formel: C₇H₁₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD05022464
CAS-Nummer: 88536-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR