1-Methyl-4-(hydroxyethyl)piperidine 98%
Lieferant: Apollo Scientific
Synonyme:
2-(1-Methylpiperidino)ethanol, 2-(1-Methylpiperidin-4-yl)ethanol
OR909465-1G
APOSOR909465-1GEA
223
EUR
APOSOR909465-1G
1-Methyl-4-(hydroxyethyl)piperidine 98%
1-Methyl-4-(hydroxyethyl)piperidine
Formel:
C₈H₁₇NO Molecular Weight: 143,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13188565 CAS-Nummer: 21156-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...