2-Methyl-2-(4-nitrophenyl)propanenitrile 95%

Lieferant: Apollo Scientific

OR909564-1G OR909564-5G
APOSOR909564-1GEA 94.4 EUR
APOSOR909564-1G APOSOR909564-5G
2-Methyl-2-(4-nitrophenyl)propanenitrile 95%
2-Methyl-2-(4-nitrophenyl)propanenitrile
Formel: C₁₀H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00844930
CAS-Nummer: 71825-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR