1-Boc-4-chlorpiperidin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 4-chloropiperidine-1-carboxylate
OR909607-1G
OR909607-25G
OR909607-5G
APOSOR909607-1GEA
109
EUR
APOSOR909607-1G
APOSOR909607-25G
APOSOR909607-5G
1-Boc-4-chlorpiperidin 97%
1-Boc-4-chlorpiperidin
Formel:
C₁₀H₁₈ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115040 CAS-Nummer: 154874-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...