4,7-Dimethyl-benzothiazol-2-ylamine 96%
Lieferant: Apollo Scientific
Synonyme:
4,7-Dimethylbenzo[d]thiazol-2-amine
OR909631-10G
OR909631-1G
OR909631-5G
APOSOR909631-10GEA
438
EUR
APOSOR909631-10G
APOSOR909631-1G
APOSOR909631-5G
4,7-Dimethyl-benzothiazol-2-ylamine 96%
4,7-Dimethyl-benzothiazol-2-ylamine
Formel:
C₉H₁₀N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971844 CAS-Nummer: 78584-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...