Ethyl-5-Isopropylpyrazole-3-carboxylate 98%

Lieferant: Apollo Scientific

Synonyme: Ethyl 3-isopropyl-1H-pyrazole-5-carboxylate

OR909670-1G OR909670-5G
APOSOR909670-1GEA 145 EUR
APOSOR909670-1G APOSOR909670-5G
Ethyl-5-Isopropylpyrazole-3-carboxylate 98%
Ethyl-5-Isopropylpyrazole-3-carboxylate
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08533270
CAS-Nummer: 78208-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR