8-Methoxy-2-tetralon
Lieferant: Apollo Scientific
Synonyme:
8-Methoxy-3,4-dihydronaphthalen-2(1H)-one
OR9097-1G
OR9097-5G
OR9097-10G
APOSOR9097-1GEA
261
EUR
APOSOR9097-1G
APOSOR9097-5G
APOSOR9097-10G
8-Methoxy-2-tetralon
8-Methoxy-2-tetralon
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,22 g/mol Schmelzpunkt: 48…55 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411220 CAS-Nummer: 5309-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...