3-Chlor-6-(cyclopropylamino)pyridazin 98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-N-cyclopropylpyridazin-3-amine

OR909779-5G
APOSOR909779-5GEA 224 EUR
APOSOR909779-5G
3-Chlor-6-(cyclopropylamino)pyridazin 98%
3-Chloro-6-(cyclopropylamino)pyridazine
Formel: C₇H₈ClN₃
Molecular Weight: 169,6100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800959
CAS-Nummer: 941294-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR