2-Chlor-6-(1-imidazolyl)pyrazin 98%
Lieferant: Apollo Scientific
OR909781-1G
OR909781-5G
APOSOR909781-1GEA
168
EUR
APOSOR909781-1G
APOSOR909781-5G
2-Chlor-6-(1-imidazolyl)pyrazin 98%
2-Chloro-6-(1-imidazolyl)pyrazine
Formel:
C₇H₅ClN₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800962 CAS-Nummer: 941294-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...