Methyl-1-methylpiperidine-4-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-methylpiperidine-4-carboxylate

OR909799-100G
APOSOR909799-100GEA 95.7 EUR
APOSOR909799-100G
Methyl-1-methylpiperidine-4-carboxylate 97%
Methyl-1-methylpiperidine-4-carboxylate
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658483
CAS-Nummer: 1690-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR