1-(3-Chlor-2-methylphenyl)pyrrolidin 97%

Lieferant: Apollo Scientific

OR909857-10G OR909857-1G OR909857-5G
APOSOR909857-10GEA 0 EUR
APOSOR909857-10G APOSOR909857-1G APOSOR909857-5G
1-(3-Chlor-2-methylphenyl)pyrrolidin 97%
1-(3-Chlor-2-methylphenyl)pyrrolidin
Formel: C₁₁H₁₄ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD09972165
CAS-Nummer: 1020253-09-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR