6-Methyl-3-isochinolinol 96%

Lieferant: Apollo Scientific

Synonyme: 6-Methyl-3-hydroxyisoquinoline

OR909923-1G OR909923-5G
APOSOR909923-1GEA 168 EUR
APOSOR909923-1G APOSOR909923-5G
6-Methyl-3-isochinolinol 96%
6-Methyl-3-isochinolinol
Formel: C₁₀H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12546459
CAS-Nummer: 51463-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR