3-(p-Tolyl)propionsäure 97%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Methylphenyl)propionsäure, 3-(4-Methylphenyl)propansäure
OR909967-100G
APOSOR909967-100GEA
206
EUR
APOSOR909967-100G
3-(p-Tolyl)propionsäure 97%
3-(p-Tolyl)propionsäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016561 CAS-Nummer: 1505-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...