N-(3-Pyridyl)pivalamid 98%
Lieferant: Apollo Scientific
Synonyme:
2,2-Dimethyl-N-pyridin-3-yl-propionamide, 2,2-Dimehtyl-N-pyridin-3-yl-propionamide
OR909970-100G
OR909970-25G
APOSOR909970-100GEA
336
EUR
APOSOR909970-100G
APOSOR909970-25G
N-(3-Pyridyl)pivalamid 98%
N-(3-Pyridyl)pivalamid
Formel:
C₁₀H₁₄N₂O Molecular Weight: 178,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00996243 CAS-Nummer: 70298-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...