4-Brom-1,3-dimethyl-1H-pyrazol 95%
Lieferant: Apollo Scientific
OR910091-1G
OR910091-25G
APOSOR910091-1GEA
0
EUR
APOSOR910091-1G
APOSOR910091-25G
4-Brom-1,3-dimethyl-1H-pyrazol 95%
4-Bromo-1,3-dimethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175,0265 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00985488 CAS-Nummer: 5775-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...