1-(2-Piperidinoethyl)-1H-pyrazol 97%

Lieferant: Apollo Scientific

OR910111-1G OR910111-250MG
APOSOR910111-1GEA 364 EUR
APOSOR910111-1G APOSOR910111-250MG
1-(2-Piperidinoethyl)-1H-pyrazol 97%
1-(2-Piperidinoethyl)-1H-pyrazol
Formel: C₁₀H₁₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03727253
CAS-Nummer: 1187385-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR