1-Benzyl-1H-pyrazol-4-carbaldehyd 95%

Lieferant: Apollo Scientific

Synonyme: 1-Benzyl-1H-pyrazole-4-carboxaldehyde

OR910197-1G OR910197-5G
APOSOR910197-1GEA 74.1 EUR
APOSOR910197-1G APOSOR910197-5G
1-Benzyl-1H-pyrazol-4-carbaldehyd 95%
1-Benzyl-1H-pyrazole-4-carbaldehyde
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,2099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179567
CAS-Nummer: 63874-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR