2-Bromothiazole-5-carbonitrile 98%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-5-cyanothiazole

OR910217-1G OR910217-5G
APOSOR910217-1GEA 112 EUR
APOSOR910217-1G APOSOR910217-5G
2-Bromothiazole-5-carbonitrile 98%
2-Bromothiazole-5-carbonitrile
Formel: C₄HBrN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09909651
CAS-Nummer: 440100-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR