1-tert-Butoxycarbonyl-1,2,4-triazole 98%
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 1H-1,2,4-triazole-1-carboxylate, 1-Boc-1,2,4-triazole
OR910221-25G
OR910221-5G
APOSOR910221-25GEA
316
EUR
APOSOR910221-25G
APOSOR910221-5G
1-tert-Butoxycarbonyl-1,2,4-triazole 98%
1-tert-Butoxycarbonyl-1,2,4-triazole
Formel:
C₇H₁₁N₃O₂ Molecular Weight: 169.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042665 CAS-Nummer: 41864-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...