2-Chloro-1-methylbenzimidazole 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-1-methyl-1H-benzo[d]imidazole

OR910286-5G
APOSOR910286-5GEA 194 EUR
APOSOR910286-5G
2-Chloro-1-methylbenzimidazole 98%
2-Chloro-1-methylbenzimidazole
Formel: C₈H₇ClN₂
Molecular Weight: 166,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00769821
CAS-Nummer: 1849-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR