4-Brom-5-chlor-2-nitroanilin 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR910289-25G
OR910289-5G
OR910289-1G
APOSOR910289-25GEA
88.6
EUR
APOSOR910289-25G
APOSOR910289-5G
APOSOR910289-1G
4-Brom-5-chlor-2-nitroanilin 95%
4-Brom-5-chlor-2-nitroanilin
Formel:
C₆H₄BrClN₂O₂ Molecular Weight: 251.4652 g/mol Schmelzpunkt: 195…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100159 CAS-Nummer: 827-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...