1-Ethyl-1H-pyrazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
OR910462-1G
OR910462-5G
APOSOR910462-1GEA
43.5
EUR
APOSOR910462-1G
APOSOR910462-5G
1-Ethyl-1H-pyrazole-5-carbaldehyde 95%
1-Ethyl-1H-pyrazole-5-carbaldehyde
Formel:
C₆H₈N₂O Molecular Weight: 124.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03130036 CAS-Nummer: 902837-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...