6,8-Dimethylchinolin-2(1H)-on 98%

Lieferant: Apollo Scientific

OR910493-1G OR910493-25G OR910493-5G
APOSOR910493-1GEA 132 EUR
APOSOR910493-1G APOSOR910493-25G APOSOR910493-5G
6,8-Dimethylchinolin-2(1H)-on 98%
6,8-Dimethylchinolin-2(1H)-on
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD15143578
CAS-Nummer: 54904-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR