3-Ethoxybenzoesäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR910911-100G
OR910911-25G
OR910911-5G
APOSOR910911-100GEA
169
EUR
APOSOR910911-100G
APOSOR910911-25G
APOSOR910911-5G
3-Ethoxybenzoesäure 99%
3-Ethoxybenzoic acid
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016517 CAS-Nummer: 621-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...