3-Phenylbenzonitril 97%
Lieferant: Apollo Scientific
Synonyme:
[1,1'-biphenyl]-3-carbonitrile
OR910931-5G
APOSOR910931-5GEA
119
EUR
APOSOR910931-5G
3-Phenylbenzonitril 97%
3-Phenylbenzonitril
Formel:
C₁₃H₉N Molecular Weight: 179,2173 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236364 CAS-Nummer: 24973-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...