3-Nitro-2-(1-pyrrolidinyl)pyridine 97%

Lieferant: Apollo Scientific

OR911068-1G OR911068-5G
APOSOR911068-1GEA 127 EUR
APOSOR911068-1G APOSOR911068-5G
3-Nitro-2-(1-pyrrolidinyl)pyridine 97%
3-Nitro-2-(1-pyrrolidinyl)pyridine
Formel: C₉H₁₁N₃O₂
Molecular Weight: 193.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001300
CAS-Nummer: 26820-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR