3-(4-Ethoxyphenyl)propionsäure 95%
Lieferant: Apollo Scientific
Synonyme:
3-(4-Ethoxyphenyl)propanoic acid
OR911154-25G
APOSOR911154-25GEA
243
EUR
APOSOR911154-25G
3-(4-Ethoxyphenyl)propionsäure 95%
3-(4-Ethoxyphenyl)propionsäure
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016557 CAS-Nummer: 4919-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...