2-Methyl-6-nitroanisol 98%
Lieferant: Apollo Scientific
OR911188-10G
OR911188-25G
OR911188-5G
APOSOR911188-10GEA
277
EUR
APOSOR911188-10G
APOSOR911188-25G
APOSOR911188-5G
2-Methyl-6-nitroanisol 98%
2-Methyl-6-nitroanisol
Formel:
C₈H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00661626 CAS-Nummer: 18102-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...