(1-Hexadecyl)tributylphosphoniumbromid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Hexadecyltri-n-butylphosphonium bromide, Tributylhexadecylphosphonium bromide, Tributyl(hexadecyl)phosphonium bromide, (n-Hexadecyl)tri-n-butylphosphonium bromide
OR9113-100G
OR9113-25G
APOSOR9113-100GEA
219
EUR
APOSOR9113-100G
APOSOR9113-25G
(1-Hexadecyl)tributylphosphoniumbromid 95+%
(1-Hexadecyl)tributylphosphoniumbromid
Formel:
C₂₈H₆₀BrP Molecular Weight: 507,65 g/mol Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011775 CAS-Nummer: 14937-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...