2,3-Dichloranisol 98%

Lieferant: Apollo Scientific

Synonyme: 1,2-Dichloro-3-methoxybenzene

OR911322-100G OR911322-500G
APOSOR911322-100GEA 66.7 EUR
APOSOR911322-100G APOSOR911322-500G
2,3-Dichloranisol 98%
2,3-Dichloranisol
Formel: C₇H₆Cl₂O
Molecular Weight: 177,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000545
CAS-Nummer: 1984-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR