L-Methioninmethylester Hydrochlorid 96%
Lieferant: Apollo Scientific
Synonyme: H-Met-OMe.HCl, H-Met-OMe·HCl
OR911324-100G
APOSOR911324-100GEA 84.1 EUR
APOSOR911324-100G
L-Methioninmethylester Hydrochlorid 96%
L-Methioninmethylester Hydrochlorid
Formel: C₆H₁₃NO₂S·HCl
Molecular Weight: 199,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012491
CAS-Nummer: 2491-18-1