Ethyl-2-ethoxybenzoat 95%
Lieferant: Apollo Scientific
Synonyme:
2-Ethoxybenzoic Acid Ethyl Ester, Ethyl 2-ethoxybenzoate
OR911336-100G
APOSOR911336-100GEA
121
EUR
APOSOR911336-100G
Ethyl-2-ethoxybenzoat 95%
Ethyl-2-ethoxybenzoate
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142907 CAS-Nummer: 6290-24-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...