Cyclohexylphenylketon 95%

Lieferant: Apollo Scientific

OR911369-100G OR911369-25G
APOSOR911369-100GEA 98.6 EUR
APOSOR911369-100G APOSOR911369-25G
Cyclohexylphenylketon 95%
Cyclohexylphenylketon
Formel: C₁₃H₁₆O
Molecular Weight: 188,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001467
CAS-Nummer: 712-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR