1-Benzyl-2,4-imidazolidindion 95%
Lieferant: Apollo Scientific
Synonyme:
1-Benzylimidazolidine-2,4-dione
OR911408-100G
OR911408-25G
APOSOR911408-100GEA
289
EUR
APOSOR911408-100G
APOSOR911408-25G
1-Benzyl-2,4-imidazolidindion 95%
1-Benzylhydantoin
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190,1986 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00466790 CAS-Nummer: 6777-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...