4-Hydroxy-6-methylnicotinic acid 98%

Lieferant: Apollo Scientific

Synonyme: 3-Carboxy-6-methyl-4-pyridone, 6-Methyl-4-oxo-1,4-dihydropyridine-3-carboxylic acid

OR911427-500G
APOSOR911427-500GEA 169 EUR
APOSOR911427-500G
4-Hydroxy-6-methylnicotinic acid 98%
4-Hydroxy-6-methylnicotinic acid
Formel: C₇H₇NO₃
Molecular Weight: 153,137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038153
CAS-Nummer: 67367-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR