2,4-Dimethylbenzaldehyd 95%
Lieferant: Apollo Scientific
OR911450-100G
OR911450-25G
APOSOR911450-100GEA
456
EUR
APOSOR911450-100G
APOSOR911450-25G
2,4-Dimethylbenzaldehyd 95%
2,4-Dimethylbenzaldehyde
Formel:
C₉H₁₀O Molecular Weight: 134,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003340 CAS-Nummer: 15764-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...