(2S)-2-Amino-4-methoxy-4-oxobutansäurehydrochlorid 98%
Lieferant: Apollo Scientific
Achtung
Synonyme: H-Asp(OMe)-OH·HCl, L-Aspartic acid 4-methyl ester hydrochloride, L-Aspartatic Acid 4-Methyl Ester Hydrochloride, L-Aspartic acid β-methyl ester hydrochloride, H-Asp(OMe)-OH.HCl, H-Asp(OMe)-OH β-Methyl L-aspartate hydrochloride
OR911506-100G OR911506-500G OR911506-25G
APOSOR911506-100GEA 94.4 EUR
APOSOR911506-100G APOSOR911506-500G APOSOR911506-25G
(2S)-2-Amino-4-methoxy-4-oxobutansäurehydrochlorid 98%
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
Formel: C₅H₉NO₄·HCl
Molecular Weight: 183.6 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00038972
CAS-Nummer: 16856-13-6