(±)-3-Chlormandelsäure 98%

Lieferant: Apollo Scientific

Synonyme: (±)-3-Chlorphenylglykolsäure, (±)-2-(3-Chlorphenyl)-2-hydroxyessigsäure, (±)-2-(3-Chlorphenyl)-2-hydroxyethansäure

OR911512-100G OR911512-25G
APOSOR911512-100GEA 237 EUR
APOSOR911512-100G APOSOR911512-25G
(±)-3-Chlormandelsäure 98%
(±)-3-Chlormandelsäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00829409
CAS-Nummer: 16273-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR