Methyl-5-methoxysalicylat 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-hydroxy-5-methoxybenzoate

OR911527-100G OR911527-25G
APOSOR911527-100GEA 340 EUR
APOSOR911527-100G APOSOR911527-25G
Methyl-5-methoxysalicylat 97%
Methyl-5-methoxysalicylat
Formel: C₉H₁₀O₄
Molecular Weight: 182,1733 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017185
CAS-Nummer: 2905-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR