6-Methoxy-o-toluidin 95%

Lieferant: Apollo Scientific

Synonyme: 1-Amino-2-methoxy-6-methylbenzol, 2-Amino-3-methylanisol, 2-Amino-3-methoxytoluol, 2-Methoxy-6-methylanilin, 6-Methyl-o-anisidin

OR911595-25G
APOSOR911595-25GEA 169 EUR
APOSOR911595-25G
6-Methoxy-o-toluidin 95%
6-Methoxy-o-toluidin
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075455
CAS-Nummer: 50868-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR