1-Phenyl-1,2-propandion 98%

Lieferant: Apollo Scientific

Synonyme: 1-Phenylpropan-1,2-dion

OR911601-100G OR911601-25G
APOSOR911601-100GEA 183 EUR
APOSOR911601-100G APOSOR911601-25G
1-Phenyl-1,2-propandion 98%
1-Phenyl-1,2-propandion
Formel: C₆H₅C(O)C(O)CH₃
Molecular Weight: 148.16 g/mol
Siedepunkt: 228
Lagertemperatur: Raumtemperatur
MDL: MFCD00008755
CAS-Nummer: 579-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR