2-Propylimidazol 95%

Lieferant: Apollo Scientific

OR911603-500G
APOSOR911603-500GEA 89.9 EUR
APOSOR911603-500G
2-Propylimidazol 95%
2-Propylimidazole
Formel: C₆H₁₀N₂
Molecular Weight: 110,1570 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059158
CAS-Nummer: 50995-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR