2,4-Dimethylchinolin 95%
Lieferant: Apollo Scientific
OR911716-5G
APOSOR911716-5GEA
231
EUR
APOSOR911716-5G
2,4-Dimethylchinolin 95%
2,4-Dimethylquinoline
Formel:
C₁₁H₁₁N Molecular Weight: 157,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006760 CAS-Nummer: 1198-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...