1-Propyl-1H-imidazol 97%

Lieferant: Apollo Scientific

Synonyme: 1-Propyl-1H-imidazole

OR911741-500G
APOSOR911741-500GEA 198 EUR
APOSOR911741-500G
1-Propyl-1H-imidazol 97%
1-Propylimidazole
Formel: C₆H₁₀N₂
Molecular Weight: 110,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060494
CAS-Nummer: 35203-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR