3-Phenylpropanamid  98%

Lieferant: Apollo Scientific

Synonyme: 3-Phenylpropanamide

OR911750-25G OR911750-5G
APOSOR911750-25GEA 241 EUR
APOSOR911750-25G APOSOR911750-5G
3-Phenylpropanamid  98%
3-Phenylpropanamid 
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00025535
CAS-Nummer: 102-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR